Få mere at vide
Trimethylsulfoxonium-d9 Iodide, TRC
Artikelnummer. 30455727
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30455727 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30455727 Leverandør Toronto Research Chemicals Leverandørnr. TRCT797502100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trimethylsulfoxonium-d9 Iodide
CAS 23726-00-3
Molekylær formel C3D9IOS
Mængde 100 mg
Synonym Tri(methyl-d3)-sulfoxonium Iodide
InChI formel InChI=1S/C3H8OS.HI.H/c1-5(2,3)4;;/h1H2,2-3H3;1H;/q+1;;/p-1/i1D2,2D3,3D3;;1+1
SMIL O=[S+](C([2H])([2H])[2H])([C]([2H])[2H])C([2H])([2H])[2H].[2H].[I-]
IUPAC navn Tri(trideuteriomethyl)sulfoxonium iodide
Molekylvægt (g/mol) 229.13
Formel vægt 229