Få mere at vide
Trimethylamine-d9 Hydrochloride, TRC
Artikelnummer. 30451045
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30451045 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451045 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79580710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trimethylamine-d9 Hydrochloride
CAS 18856-86-5
Renhedsgrad noter HPLC
Molekylær formel C3 2H9 N . Cl H
Mængde 10 mg
Synonym Methan-d3-amine','N,N-di(methyl-d3)-','hydrochloride (9CI), Tri(methyl-d3)amine','hydrochloride (8CI), N,N-Di(methyl-d3)methan-d3-amine hydrochloride, Trimethylamine-d9 hydrochloride, Tri(methyl-d3)ammonium chloride, Trimethylammonium-d9 hydrochloride, Trimethylamine-D9 hydrochloride
InChI formel InChI=1S/C3H9N.ClH/c1-4(2)3;/h1-3H3;1H/i1D3,2D3,3D3;
SMIL Cl.[2H]C([2H])([2H])N(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,1,1-trideuterio-N,N-bis(trideuteriomethyl)methanamine;hydrochloride
Molekylvægt (g/mol) 104.63
Formel vægt 104.1067
Procent renhed >95