missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetylacetonitrile, 99%, Thermo Scientific Chemicals

Catalog Number p-7023176
360.69 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
59997-51-2
C7H11NO
125.171
MFCD00010208
MXZMACXOMZKYHJ-UHFFFAOYSA-N
pivaloylacetonitrile, trimethylacetylacetonitrile, 4,4-dimethyl-3-oxovaleronitrile, pentanenitrile, 4,4-dimethyl-3-oxo, 4,4-dimethyl-3-oxo-pentanenitrile, cyanopinacolone, pivaloyl acetonitrile, 1-cyano-3,3-dimethyl-2-butanone, valeronitrile, 4,4-dimethyl-3-oxo, pyvaolylacetonitrile
108871
4,4-dimethyl-3-oxopentanenitrile
CC(C)(C)C(=O)CC#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
59997-51-2
C7H11NO
125.171
MFCD00010208
MXZMACXOMZKYHJ-UHFFFAOYSA-N
pivaloylacetonitrile, trimethylacetylacetonitrile, 4,4-dimethyl-3-oxovaleronitrile, pentanenitrile, 4,4-dimethyl-3-oxo, 4,4-dimethyl-3-oxo-pentanenitrile, cyanopinacolone, pivaloyl acetonitrile, 1-cyano-3,3-dimethyl-2-butanone, valeronitrile, 4,4-dimethyl-3-oxo, pyvaolylacetonitrile
108871
4,4-dimethyl-3-oxopentanenitrile
CC(C)(C)C(=O)CC#N

Trimethylacetylacetonitrile is a useful intermediate. 4,4-Dimethyl-3-oxopentanenitrile has been used in the preparation of 4,4-dimethyl-3-oxo-2-benzylpentanenitrile, 4,4-dimethyl-3-oxo-2-(4-hydroxybenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(4-methoxybenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(4-fluorobenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(3,4-methylenedioxybenzyl)pentanenitrile, 1-(5-tert-butyl-2-p-tolyl-2H-pyrazol-3-yl)-3-phenylurea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetylacetonitrile is a useful intermediate. 4,4-Dimethyl-3-oxopentanenitrile has been used in the preparation of 4,4-dimethyl-3-oxo-2-benzylpentanenitrile, 4,4-dimethyl-3-oxo-2-(4-hydroxybenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(4-methoxybenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(4-fluorobenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(3,4-methylenedioxybenzyl)pentanenitrile, 1-(5-tert-butyl-2-p-tolyl-2H-pyrazol-3-yl)-3-phenylurea.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY
Melting Point 66°C to 70°C
Quantity 25 g
UN Number UN3439
Beilstein 1746674
Solubility Information Insoluble in water.
Formula Weight 125.17
Percent Purity 99%
Chemical Name or Material Trimethylacetylacetonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'