missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl phosphate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7080865
Click to view available options
Quantity:
500 g
2500 g
This item is not returnable. View return policy
512-56-1
C3H9O4P
140.08
MFCD00008348
WVLBCYQITXONBZ-UHFFFAOYSA-N
trimethylphosphate, phosphoric acid, trimethyl ester, trimethyl orthophosphate, tmpo, trimethoxyphosphine oxide, phosphoric acid trimethyl ester, o,o,o-trimethyl phosphate, trimethylfosfat, phosphate, trimethyl, trimethylfosfat czech
10541
CHEBI:46324
trimethyl phosphate
COP(=O)(OC)OC
This item is not returnable. View return policy
512-56-1
C3H9O4P
140.08
MFCD00008348
WVLBCYQITXONBZ-UHFFFAOYSA-N
trimethylphosphate, phosphoric acid, trimethyl ester, trimethyl orthophosphate, tmpo, trimethoxyphosphine oxide, phosphoric acid trimethyl ester, o,o,o-trimethyl phosphate, trimethylfosfat, phosphate, trimethyl, trimethylfosfat czech
10541
CHEBI:46324
trimethyl phosphate
COP(=O)(OC)OC

Trimethyl phosphate is used as a color inhibitor for polyester and other polymer preparations, flame retardants and plasticizer. It acts as a solvent for aromatic halogenations and nitration reactions. It is used as a mild methylating reagent which is used in the dimethylation of aniline and related heterocyclic compound. Furthermore, it is involved in the synthesis of 1-methyl-pyridinium; dimethyl phosphate by reacting with pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethyl phosphate is used as a color inhibitor for polyester and other polymer preparations, flame retardants and plasticizer. It acts as a solvent for aromatic halogenations and nitration reactions. It is used as a mild methylating reagent which is used in the dimethylation of aniline and related heterocyclic compound. Furthermore, it is involved in the synthesis of 1-methyl-pyridinium; dimethyl phosphate by reacting with pyridine.

Solubility
Miscible with water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -46°C
Density 1.213
Boiling Point 196°C to 198°C
Flash Point >150°C (302°F)
Odor Pleasant
Linear Formula (CH3O)3PO
Refractive Index 1.397
Quantity 500 g
Beilstein 1071731
Sensitivity Moisture sensitive
Solubility Information Miscible with water.
Formula Weight 140.08
Percent Purity ≥98%
Chemical Name or Material Trimethyl phosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'