Få mere at vide
Trimethyl-d9-sulfonium Iodide, TRC
Artikelnummer. 30456066
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30456066 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30456066 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79748150MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trimethyl-d9-sulfonium Iodide
CAS 106776-17-4
Renhedsgrad noter HPLC
Molekylær formel C3 D9 S . I
Mængde 50 mg
Synonym Tri(methyl-d3)sulfonium Iodide, Sulfonium','tri(methyl-d3)-','iodide (9CI)
InChI formel InChI=1S/C3H9S.HI/c1-4(2)3;/h1-3H3;1H/q+1;/p-1/i1D3,2D3,3D3;
SMIL [I-].[2H]C([2H])([2H])[S+](C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn tris(trideuteriomethyl)sulfanium;iodide
Molekylvægt (g/mol) 213.1285
Formel vægt 213.0035
Procent renhed >95