Få mere at vide
Trimethyl-d9-sulfonium Iodide, CDN
Artikelnummer. 30312488
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30312488 0.5 g 0.5g
30312333 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312488 Leverandør CDN Leverandørnr. CDND60930.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trimethyl-d9-sulfonium Iodide
CAS 106776-17-4
Molekylær formel C3 D9 S . I
Mængde 0.5 g
Synonym Tri(methyl-d3)sulfonium Iodide, Sulfonium, tri(methyl-d3)-, iodide (9 CI)
InChI formel InChI=1 S/C3H9S.HI/c1-4(2)3;/h1-3H3;1 H/q+1;/p-1/i1D3,2D3,3D3;
SMIL [I-].[2 H]C([2 H])([2 H])[S+](C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn tris(trideuteriomethyl)sulfanium;iodide
Molekylvægt (g/mol) 213.1285
Formel vægt 213.0035 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity