missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl borate, 99.9995+% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049809
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
121-43-7
C3H9BO3
103.912
MFCD00008346
WRECIMRULFAWHA-UHFFFAOYSA-N
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
8470
CHEBI:38913
trimethyl borate
B(OC)(OC)OC
This item is not returnable. View return policy
121-43-7
C3H9BO3
103.912
MFCD00008346
WRECIMRULFAWHA-UHFFFAOYSA-N
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
8470
CHEBI:38913
trimethyl borate
B(OC)(OC)OC

Trimethyl borate is a useful reagent in organic synthesis. It is involved in the production of resins, waxes and paints and acts as a methylation agent. As a boron source, it is used to prepare flame retardants, anti-oxidants and corrosion inhibitors. It reacts with Grignard reagents followed by hydrolysis to prepare boronic acid. It is also used as a precursor of borate esters, which finds application in the Suzuki coupling reaction. It is an intermediate in the preparation of sodium borohydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.915g/mL
Assay Percent Range >99.9995% (metals basis)
Linear Formula (CH3O)3B
Quantity 10 g
UN Number UN2416
Beilstein 1697939
Merck Index 14,9712
Formula Weight 103.92
Percent Purity ≥99.9995%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Trimethyl borate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'