Få mere at vide
Trimethoprim-d9 (Major), TRC
Artikelnummer. 30454635
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30454635 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454635 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7956171MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trimethoprim D9 (trimethoxy D9)
CAS 1189460-62-5
Renhedsgrad noter HPLC
Molekylær formel C14 D9 H9 N4 O3
Mængde 1 mg
Synonym Trimethoprim D9 (trimethoxy D9), 2,4-Pyrimidinediamine','5-[[3,4,5-tri(methoxy-d3)phenyl]methyl]-, 5-[[3,4,5-Tris(trideuteriomethoxy)phenyl]methyl]pyrimidine-2,4-diamine, Trimethoprim D9 (trimethoxy D9), 2,4-Pyrimidinediamine','5-[[3,4,5-tri(methoxy-d3)phenyl]methyl]-, 5-[[3,4,5-Tris(trideuteriomethoxy)phenyl]methyl]pyrimidine-2,4-diamine
InChI formel InChI=1S/C14H18N4O3/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15/h5-7H,4H2,1-3H3,(H4,15,16,17,18)/i1D3,2D3,3D3
SMIL [2H]C([2H])([2H])Oc1cc(Cc2cnc(N)nc2N)cc(OC([2H])([2H])[2H])c1OC([2H])([2H])[2H]
IUPAC navn 5-[[3,4,5-tris(trideuteriomethoxy)phenyl]methyl]pyrimidine-2,4-diamine
Molekylvægt (g/mol) 299.37
Formel vægt 299.1944
Procent renhed >95