Få mere at vide
Trimethoprim-d3 (Major), TRC
Artikelnummer. 30449033
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30449033 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449033 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7956232.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trimethoprim-d3
CAS 1189923-38-3
Molekylær formel C14 D5 H15 N4 O3
Mængde 2.5 mg
Synonym Trimethoprim D3 (4-methoxy D3)
InChI formel InChI=1S/C14H18N4O3.2H/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15;;/h5-7H,4H2,1-3H3,(H4,15,16,17,18);;/i3D3;2*1+1
SMIL [2H].[2H].[2H]C([2H])([2H])Oc1c(OC)cc(Cc2cnc(N)nc2N)cc1OC
IUPAC navn 5-[[3,5-dimethoxy-4-(trideuteriomethoxy)phenyl]methyl]pyrimidine-2,4-diamine
Molekylvægt (g/mol) 297.364
Formel vægt 297.185