Få mere at vide
Trimethoprim 3-N-Oxide, TRC
Artikelnummer. 30134796
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134796 10 mg
17953155 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134796 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79754510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27653-67-4
Molekylær formel C14 H18 N4 O4
Molekylvægt (g/mol) 306.3171
Synonym Trimethoprim 3-oxide, 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine 3-Oxide
IUPAC navn 3-oxido-5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidin-3-ium-2,4-diamine
SMIL COc1cc(Cc2cnc(N)[n+]([O-])c2N)cc(OC)c1OC
Tekniske data
Kemisk navn eller materiale Trimethoprim 3-N-Oxide
Mængde 10 mg
InChI formel InChI=1S/C14H18N4O4/c1-20-10-5-8(6-11(21-2)12(10)22-3)4-9-7-17-14(16)18(19)13(9)15/h5-7H,4,15H2,1-3H3,(H2,16,17)
Formel vægt 306.1328