Få mere at vide
Trimethoprim-13C3, TRC
Artikelnummer. 17997434
Skift visning
Mængde:
0.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17997434 5 mg -
17948404 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17997434 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7956165MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189970-95-3
Molekylær formel 13C3 C11 H18 N4 O3
Molekylvægt (g/mol) 293.3
Synonym Trimethoprim (pyrimidine-4,5,6-13C3)
IUPAC navn 5-[(3,4,5-trimethoxyphenyl)methyl](4,5,6-^{13}C_{3})pyrimidine-2,4-diamine
SMIL COc1cc(C[13c]2[13cH]nc(N)n[13c]2N)cc(OC)c1OC
Tekniske data
Kemisk navn eller materiale Trimethoprim-13C3
Mængde 5 mg
InChI formel InChI=1S/C14H18N4O3/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15/h5-7H,4H2,1-3H3,(H4,15,16,17,18)/i7+1,9+1,13+1
Formel vægt 293.148