Få mere at vide
Trimetazidine Dihydrochloride, Mikromol™
Beskrivelse
Trimetazidine Dihydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Trimetazidine Dihydrochloride |
| CAS | 13171-25-0 |
| Analyt- eller komponentnavne | Trimetazidine Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H22 N2 O3 . 2 Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Trimetazidine Dihydrochloride, Piperazine, 1-[(2,3,4-trimethoxyphenyl)methyl]-, dihydrochloride (9CI), Piperazine, 1-(2,3,4-trimethoxybenzyl)-, dihydrochloride (7CI,8CI), 1-(2,3,4-Trimethoxybenzyl)piperazine dihydrochloride, Kyurinett, S 4004, Vastarel, Vastarel F, Yoshimilon |
| InChI formel | InChI=1S/C14H22N2O3.2ClH/c1-17-12-5-4-11(13(18-2)14(12)19-3)10-16-8-6-15-7-9-16;;/h4-5,15H,6-10H2,1-3H3;2*1H |
| SMIL | Cl.Cl.COc1ccc(CN2CCNCC2)c(OC)c1OC |
| IUPAC navn | 1-[(2,3,4-trimethoxyphenyl)methyl]piperazine;dihydrochloride |
| Vis mere |