Få mere at vide
Trimetaphan Camsilate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Trimetaphan Camsilate |
| CAS | 68-91-7 |
| Analyt- eller komponentnavne | Trimetaphan Camsilate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C22 H25 N2 O S . C10 H15 O4 S |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Decahydro-2-oxo-1,3-bis(phenylmethyl)thieno[1',2':1,2]thieno[3,4-d]imidazol-5-ium salt with (1S)- 7,7-dimethyl-2-oxobicyclo[2.2.1]heptane-1-methanesulfonic acid, Trimethaphan camphorsulfonate, Trimetaphan camsylate, 1,3-Dibenzyldecahydro-2-oxoimidazo[4,5-c]thieno[1,2-a]thiolium (+)-10-camphorsulfonate, 4,6-Dibenzyl-5-oxo-1-thia-4,6-diazatricyclo[6.3.0.03,7]undecanium (+)-beta-camphorsulfonate, Arfonad, Arphonad, Methioplegium, Nu 2222, Ro 2-2222, Trimetaphan camphorsulfonate, Trimetaphan camsilate, Trimetaphan camsylate, Trimethaphan 10-camphorsulfonate, Trimethaphan camsylate |
| InChI formel | InChI=1S/C22H25N2OS.C10H16O4S/c25-22-23(14-17-8-3-1-4-9-17)19-16-26-13-7-12-20(26)21(19)24(22)15-18-10-5-2-6-11-18;1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h1-6,8-11,19-21H,7,12-16H2;7H,3-6H2,1-2H3,(H,12,13,14)/q+1;/p-1 |
| Løsningstype | Neat |
| SMIL | CC1(C)C2CCC1(CS(=O)(=O)[O-])C(=O)C2.O=C3N(Cc4ccccc4)C5C[S+]6CCCC6C5N3Cc7ccccc7 |
| Vis mere |