missing translation for 'onlineSavingsMsg'
Learn More

Trimesitylphosphine, 98%, Thermo Scientific Chemicals

Catalog Number p-7025569
Click to view available options
Quantity:
1 g
5 g
23897-15-6
C27H33P
388.535
MFCD00014912
IDXDWPWXHTXJMZ-UHFFFAOYSA-N
trimesitylphosphine, tris 2,4,6-trimethylphenyl phosphine, tris 2,4,6-trimethylphenyl phosphane, phosphine, tris 2,4,6-trimethylphenyl, pubchem6447, acmc-1cs30, phosphine,tris 2,4,6-trimethylphenyl
2733417
tris(2,4,6-trimethylphenyl)phosphane
CC1=CC(=C(C(=C1)C)P(C2=C(C=C(C=C2C)C)C)C3=C(C=C(C=C3C)C)C)C
23897-15-6
C27H33P
388.535
MFCD00014912
IDXDWPWXHTXJMZ-UHFFFAOYSA-N
trimesitylphosphine, tris 2,4,6-trimethylphenyl phosphine, tris 2,4,6-trimethylphenyl phosphane, phosphine, tris 2,4,6-trimethylphenyl, pubchem6447, acmc-1cs30, phosphine,tris 2,4,6-trimethylphenyl
2733417
tris(2,4,6-trimethylphenyl)phosphane
CC1=CC(=C(C(=C1)C)P(C2=C(C=C(C=C2C)C)C)C3=C(C=C(C=C3C)C)C)C

It is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 185°C to 188°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 388.54
Percent Purity 98%
Chemical Name or Material Trimesitylphosphine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'