Få mere at vide
Trimeprazine Tartrate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Alimemazine tartrate |
| CAS | 4330-99-8 |
| Analyt- eller komponentnavne | Alimemazine tartrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | 2 C18 H22 N2 S . C4 H6 O6 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Phenothiazine, 10-[3-(dimethylamino)-2-methylpropyl]-, tartrate (2:1) (8CI), 10-(3-Dimethylamino-2-methylpropyl)phenothiazine tartrate, Alimemazine tartrate, NSC 17475, Panectyl, Repeltin, Temaril, Temaril 16, Theralene, Trimeprazine Tartrate, Vallergan, Vanectyl, 10H-Phenothiazine-10-propanamine, N,N,beta-trimethyl-, (2R,3R)-2,3-dihydroxybutanedioate (2:1), 10H-Phenothiazine-10-propanamine, N,N,beta-trimethyl-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (2:1) |
| InChI formel | InChI=1S/2C18H22N2S.C4H6O6/c2*1-14(12-19(2)3)13-20-15-8-4-6-10-17(15)21-18-11-7-5-9-16(18)20;5-1(3(7)8)2(6)4(9)10/h2*4-11,14H,12-13H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1 |
| Løsningstype | Neat |
| SMIL | CC(CN(C)C)CN1c2ccccc2Sc3ccccc13.CC(CN(C)C)CN4c5ccccc5Sc6ccccc46.O[C@H]([C@@H](O)C(=O)O)C(=O)O |
| Vis mere |