Learn More
Triisopropyl Phosphate, TRC
Product Code. 30453432
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453432 1 g -
30454167 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30453432 Supplier Toronto Research Chemicals Supplier No. TRCT7954161G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 513-02-0
Molecular Formula C9 H21 O4 P
Molecular Weight (g/mol) 224.23
Synonym Isopropyl phosphate ((C3H7O)3PO) (6CI,7CI), Phosphoric acid, triisopropyl ester (8CI), NSC 46370, NSC 62275, Triisopropoxyphosphine oxide, Triisopropyl phosphate
IUPAC Name tripropan-2-yl phosphate
SMILES CC(C)OP(=O)(OC(C)C)OC(C)C
Specifications
Quantity 1 g
InChI Formula InChI=1S/C9H21O4P/c1-7(2)11-14(10,12-8(3)4)13-9(5)6/h7-9H,1-6H3
Formula Weight 224.1177
Chemical Name or Material Triisopropyl phosphate