Få mere at vide
Trihexyphenidyl Hydrochloride, Mikromol™
Beskrivelse
Trihexyphenidyl Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Benzhexol hydrochloride |
| CAS | 52-49-3 |
| Analyt- eller komponentnavne | Benzhexol hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H31 N O . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Trihexyphenidyl Hydrochloride, alpha-Cyclohexyl-alpha-phenyl-1-piperidinepropanol hydrochloride), (+/-)-Trihexyphenidyl hydrochloride, alpha-Cyclohexyl-alpha-phenyl-1-piperidinepropanol hydrochloride, 1-Phenyl-1-cyclohexyl-3-piperidyl-1-propanol hydrochloride, 3-(1-Piperidyl)-1-cyclohexyl-1-phenyl-1-propanol hydrochloride, 3-(1-Piperidyl)-1-phenyl-1-cyclohexyl-1-propanol hydrochloride, Alpha-Cyclohexyl-alpha-phenyl-1-piperidinepropanol hydrochloride, Antitrem, Aparkane, Artane, Artane hydrochloride, Benzhexol chloride, Benzhexol hydrochloride, Broflex, Cyclodol, Pacitane, Paralest, Parcopane, Pargitan, Parkidyl, Parkinane, Parkinol, Parkinsan, Parkisan, Parkopan, Partigan, Peragit, Pipanol, Romparkin, Sedrina, Tremin, Trihexy, Trihexyphenidyl chlorhydrate, Trihexyphenidyl hydrochloride, Triphedinon, Triphenidyl, Tsiklodol |
| InChI formel | InChI=1S/C20H31NO.ClH/c22-20(18-10-4-1-5-11-18,19-12-6-2-7-13-19)14-17-21-15-8-3-9-16-21;/h1,4-5,10-11,19,22H,2-3,6-9,12-17H2;1H |
| SMIL | Cl.OC(CCN1CCCCC1)(C2CCCCC2)c3ccccc3 |
| IUPAC navn | 1-cyclohexyl-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride |
| Vis mere |