Få mere at vide
Triglyme-d6, TRC
Artikelnummer. 30454518
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30454518 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454518 Leverandør Toronto Research Chemicals Leverandørnr. TRCT793802100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Triglyme-d6
CAS 1219192-16-1
Molekylær formel C8H12D6O4
Mængde 100 mg
Synonym 2,5,8,11-Tetraoxadodecane-d6, 1,2-Bis(2-methoxyethoxy)ethane-d6, Ansul Ether 161-d6, DMTG-d6, Glyme 4, Hisolve MTM-d6, Methyltriglyme-d6, NSC 66400-d6, Triethylene Glycol Dimethyl-d6 Ether
InChI formel InChI=1S/C8H18O4/c1-9-3-5-11-7-8-12-6-4-10-2/h3-8H2,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])OCCOCCOCCOC([2H])([2H])[2H]
IUPAC navn 2,5,8,11-tetraoxadodecane-1,1,1,12,12,12-d6
Molekylvægt (g/mol) 184.26
Formel vægt 184.16