Få mere at vide
Trifluralin-(dipropyl-d14), TRC
Artikelnummer. 30451109
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30451109 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451109 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79355210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trifluralin D14 (di-n-propyl D14)
CAS 347841-79-6
Renhedsgrad noter HPLC
Molekylær formel C13 2H14 H2 F3 N3 O4
Mængde 10 mg
Synonym Benzenamine','2,6-dinitro-N,N-di(propyl-d7)-4-(trifluoromethyl)- (9CI), 2,6-Dinitro-N,N-di(propyl-d7)-4-(trifluoromethyl)benzenamine, 2,6-Dinitro-N,N-di(propyl-d7)-4-(trifluoromethyl)aniline, Trifluralin-d14, Trifluralin D14 (di-n-propyl D14), Trifluralin-D14 (N,N-dipropyl-D14)
InChI formel InChI=1S/C13H16F3N3O4/c1-3-5-17(6-4-2)12-10(18(20)21)7-9(13(14,15)16)8-11(12)19(22)23/h7-8H,3-6H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])N(c1c(cc(cc1[N+](=O)[O-])C(F)(F)F)[N+](=O)[O-])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn N,N-bis(1,1,2,2,3,3,3-heptadeuteriopropyl)-2,6-dinitro-4-(trifluoromethyl)aniline
Molekylvægt (g/mol) 349.37
Formel vægt 349.1972
Procent renhed >95