Få mere at vide
Trifluoperazine Sulfoxide N1,N4-Dioxide (Trifluoperazine N1,N4,S-Trioxide), Mikromol™
Beskrivelse
Trifluoperazine Sulfoxide N1,N4-Dioxide (Trifluoperazine N1,N4,S-Trioxide)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Trifluoperazine Sulfoxide N1,N4-Dioxide |
| CAS | 150111-58-3 |
| Analyt- eller komponentnavne | Trifluoperazine Sulfoxide N1,N4-Dioxide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H24 F3 N3 O3 S |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 10-[3-(4-Methyl-1,4-dioxido-1-piperazinyl)propyl]-2-(trifluoromethyl)-10H-phenothiazine 5-Oxide, 10-[3-(4-Methyl-1,4-dioxido-1-piperazinyl)propyl]-2-(trifluoromethyl)-10H-phenothiazine N,N',5-Trioxide, Trifluoperazine N1',N4',S-Trioxide, Trifluoperazine N1,N4,S-Trioxide, Trifluoperazine Sulfoxide N1,N4-Dioxide |
| InChI formel | InChI=1S/C21H24F3N3O3S/c1-26(28)11-13-27(29,14-12-26)10-4-9-25-17-5-2-3-6-19(17)31(30)20-8-7-16(15-18(20)25)21(22,23)24/h2-3,5-8,15H,4,9-14H2,1H3 |
| SMIL | C[N+]1([O-])CC[N+]([O-])(CCCN2c3ccccc3S(=O)c4ccc(cc24)C(F)(F)F)CC1 |
| IUPAC navn | 10-[3-(4-methyl-1,4-dioxidopiperazine-1,4-diium-1-yl)propyl]-2-(trifluoromethyl)phenothiazine 5-oxide |
| Vis mere |