Få mere at vide
Trifluoperazine Sulfoxide, Mikromol™
Beskrivelse
Trifluoperazine Sulfoxide
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Trifluoperazine Sulfoxide |
| CAS | 1549-88-8 |
| Analyt- eller komponentnavne | Trifluoperazine Sulfoxide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H24 F3 N3 O S |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 10-[3-(4-Methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-10H-phenothiazine 5-Oxide, 10-[3-(4-Methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)phenothiazine 5-Oxide, Trifluoperazine S-oxide, Trifluoperazine sulfoxide, Trifluoperazine sulphoxide, Trifluoroperazine sulfoxide, Triphthazine sulfoxide, Stelazine sulphoxide, SKF-5806 |
| InChI formel | InChI=1S/C21H24F3N3OS/c1-25-11-13-26(14-12-25)9-4-10-27-17-5-2-3-6-19(17)29(28)20-8-7-16(15-18(20)27)21(22,23)24/h2-3,5-8,15H,4,9-14H2,1H3 |
| SMIL | CN1CCN(CCCN2c3ccccc3S(=O)c4ccc(cc24)C(F)(F)F)CC1 |
| IUPAC navn | 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine 5-oxide |
| Vis mere |