Få mere at vide
Trifluoperazine N1,N4-Dioxide, Mikromol™
Artikelnummer. 30050923
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050923 100 mg
30049644 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050923 Leverandør Mikromol Leverandørnr. MM0441.11
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Trifluoperazine N1,N4-Dioxide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trifluoperazine N1,N4-Dioxide
CAS 150111-57-2
Analyt- eller komponentnavne Trifluoperazine N1,N4-Dioxide
Forsendelsestilstand Room Temperature
Molekylær formel C21 H24 F3 N3 O2 S
Certificeringer/compliance ISO 9001
Synonym 10-[3-(4-methyl-1,4-dioxido-1-piperazinyl)propyl]-2-(trifluoromethyl)-10H-Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-10H-phenothiazine N,N'-Dioxide, Trifluoperazine N1,N4-Dioxide
InChI formel InChI=1S/C21H24F3N3O2S/c1-26(28)11-13-27(29,14-12-26)10-4-9-25-17-5-2-3-6-19(17)30-20-8-7-16(15-18(20)25)21(22,23)24/h2-3,5-8,15H,4,9-14H2,1H3
SMIL C[N+]1([O-])CC[N+]([O-])(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1
IUPAC navn 10-[3-(4-methyl-1,4-dioxidopiperazine-1,4-diium-1-yl)propyl]-2-(trifluoromethyl)phenothiazine
Molekylvægt (g/mol) 439.49
Mængde 100 mg
Formel vægt 439.1541 g/mol