Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Trifloxysulfuron-sodium |
| CAS | 199119-58-9 |
| CAS Max % | ≤ 100% |
| Tæthed | 1.63 g/cm3 (20°C) |
| Farve | White |
| pH | 8.9 |
| Smeltepunkt | 168°C |
| Synonym | Sodium (4,6-dimethoxypyrimidin-2-yl)({[3-(2,2,2-trifluoroethoxy)pyridin-2-yl]sulfonyl}carbamoyl)azanide |
| Molekylvægt (g/mol) | 459.33 g/mol |
| Mængde | 100 mg |
| Vis mere |