missing translation for 'onlineSavingsMsg'
Learn More

Triethylene glycol, 99%, Thermo Scientific Chemicals

Catalog Number p-7080996
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
112-27-6
C6H14O4
150.17
MFCD00002880,MFCD00081839,MFCD01779596,MFCD01779599,MFCD01779601,MFCD01779603,MFCD01779605,MFCD01779609,MFCD01779611,MFCD01779612,MFCD01779614,MFCD01779615,MFCD01779616
ZIBGPFATKBEMQZ-UHFFFAOYSA-N
triethylene glycol, triglycol, 2,2'-ethane-1,2-diylbis oxy diethanol, trigen, triethylenglykol, triethyleneglycol, 2,2'-ethylenedioxydiethanol, 1,2-bis 2-hydroxyethoxy ethane, 3,6-dioxaoctane-1,8-diol, 2,2'-ethylenedioxy diethanol
8172
CHEBI:44926
2-[2-(2-hydroxyethoxy)ethoxy]ethanol
OCCOCCOCCO
112-27-6
C6H14O4
150.17
MFCD00002880,MFCD00081839,MFCD01779596,MFCD01779599,MFCD01779601,MFCD01779603,MFCD01779605,MFCD01779609,MFCD01779611,MFCD01779612,MFCD01779614,MFCD01779615,MFCD01779616
ZIBGPFATKBEMQZ-UHFFFAOYSA-N
triethylene glycol, triglycol, 2,2'-ethane-1,2-diylbis oxy diethanol, trigen, triethylenglykol, triethyleneglycol, 2,2'-ethylenedioxydiethanol, 1,2-bis 2-hydroxyethoxy ethane, 3,6-dioxaoctane-1,8-diol, 2,2'-ethylenedioxy diethanol
8172
CHEBI:44926
2-[2-(2-hydroxyethoxy)ethoxy]ethanol
OCCOCCOCCO

Triethylene glycol is used as a plasticizer, as an additive for hydraulic fluids and brake fluids, and as a disinfectant. It is an active component of certain pigments, printing dyes, inks and paste. It finds application as a liquid desiccant and used in the dehydration of natural gas, carbon dioxide, hydrogen sulfide and air conditioning systems. It plays as an important role in anti-freeze and de-icing products, cleaning and furnishing care products, lubricant and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylene glycol is used as a plasticizer, as an additive for hydraulic fluids and brake fluids, and as a disinfectant. It is an active component of certain pigments, printing dyes, inks and paste. It finds application as a liquid desiccant and used in the dehydration of natural gas, carbon dioxide, hydrogen sulfide and air conditioning systems. It plays as an important role in anti-freeze and de-icing products, cleaning and furnishing care products, lubricant and greases.

Solubility
Miscible with water, ethanol, acetone, benzene, toluene, acetic acid, glycerine, pyridine and aldehydes. Slightly miscible with diethyl ether and chloroform. Immiscible with oil, fat, petroleum ether and most hydrocarbons.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY
Melting Point °C to 7°C
Density 1.125 g/mL
Boiling Point 284°C to 288°C
Flash Point 165°C (329°F)
Linear Formula H(OCH2CH2)3OH
Refractive Index 1.4555
Quantity 500 mL
Beilstein 969357
Sensitivity Hygroscopic
Merck Index 14,9670
Solubility Information Miscible with water,ethanol,acetone,benzene,toluene,acetic acid,glycerine,pyridine and aldehydes. Slightly miscible with diethyl ether and chloroform. Immiscible with oil,fat,petroleum ether and most hydrocarbons.
Formula Weight 150.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Triethylene glycol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'