missing translation for 'onlineSavingsMsg'
Få mere at vide

Triethylammonium Sulfate, TRC

Product Code. 30455772
Change view
Click to view available options
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30455772 100 mg
30456889 1 g
30454745 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455772 Supplier Toronto Research Chemicals Supplier No. TRCT797470100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54272-29-6
Molekylær formel C6H15N .xH2O4S
Molekylvægt (g/mol) 199.27
Synonym N,N-diethylethanamine sulfate
IUPAC navn N,N-diethylethanamine;sulfuric acid
SMIL CCN(CC)CC.OS(=O)(=O)O

Specifications

Kemisk navn eller materiale Triethylammonium Sulfate
Mængde 100 mg
InChI formel InChI=1S/C6H15N.H2O4S/c1-4-7(5-2)6-3;1-5(2,3)4/h4-6H2,1-3H3;(H2,1,2,3,4)
Formel vægt 199.0878
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.