Få mere at vide
Triethylamine-d15, TRC
Artikelnummer. 30450928
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30450928 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30450928 Leverandør Toronto Research Chemicals Leverandørnr. TRCT797473250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Triethylamine-d15
CAS 66688-79-7
Molekylær formel C6 2H15 N
Mængde 250 mg
Synonym Ethan-1,1,2,2,2-d5-amine','N,N-di(ethyl-1,1,2,2,2-d5)- (ACI), Ethan-d5-amine','N,N-di(ethyl-d5)- (9CI), N,N-Di(ethyl-1,1,2,2,2-d5)ethan-1,1,2,2,2-d5-amine (ACI), Triethylamine-d15, Tris(pentadeuteroethyl)amine, Triethylamine-D15
InChI formel InChI=1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2
SMIL [2H]C([2H])([2H])C([2H])([2H])N(C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 1,1,2,2,2-pentadeuterio-N,N-bis(1,1,2,2,2-pentadeuterioethyl)ethanamine
Molekylvægt (g/mol) 116.282
Formel vægt 116.215