Få mere at vide
Triethyl phosphite 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Triethyl Phosphite |
| CAS | 122-52-1 |
| Analyt- eller komponentnavne | Triethyl Phosphite |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C6 H15 O3 P |
| Koncentration | 100 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Ethyl phosphite, (EtO)3P (7CI), Ethyl phosphite, Et3PO3 (4CI), NSC 5284, Triethoxyphosphine, Tris(ethoxy)phosphine, Phosphorous Acid Triethyl Ester |
| InChI formel | InChI=1S/C6H15O3P/c1-4-7-10(8-5-2)9-6-3/h4-6H2,1-3H3 |
| SMIL | CCOP(OCC)OCC |
| Vis mere |