Få mere at vide
Triethyl phosphate D15, Dr. Ehrenstorfer
Artikelnummer. 30324884
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30324884 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324884 Leverandør Dr. Ehrenstorfer Leverandørnr. DRECA17835510
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Triethyl phosphate-d15
CAS 135942-11-9
Molekylær formel C6 D15 O4 P
Mængde 25 mg
InChI formel InChI=1S/C6H15O4P/c1-4-8-11(7,9-5-2)10-6-3/h4-6H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2
SMIL [2H]C([2H])([2H])C([2H])([2H])OP(=O)(OC([2H])([2H])C([2H])([2H])[2H])OC([2H])([2H])C([2H])([2H])[2H]
IUPAC navn tris(1,1,2,2,2-pentadeuterioethyl) phosphate
Molekylvægt (g/mol) 197.25
Formel vægt 197.165 g/mol