missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyl orthoformate, ≥98.0% (GC), Honeywell™
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Melting Point | -76°C (literature) |
Density | 0.891 g/mL at 25 °C(lit.) |
Boiling Point | 143°C to 145°C |
Flash Point | 30°C |
Packaging | Glass Bottle |
Linear Formula | CH(OC2H5)3 |
Quantity | 1 L |
UN Number | UN2524 |
Beilstein | 605384 |
Percent Purity | >98% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'