missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethanolamine, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
Specifications
Melting Point | 21°C |
Color | Yellow |
pH | 10.5 |
Boiling Point | 360°C |
Packaging | Amber glass bottle |
Quantity | 2.5 L |
Vapor Density | 5.14 |
Formula Weight | 149.19g/mol |
Vapor Pressure | <0.01mmHg at 20°C |
Physical Form | Viscous Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'