missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethanolamine, ∽99%, MP Biomedicals™
Click to view available options
Quantity:
1 kg
Specifications
Color | White |
Assay Percent Range | ∽ 99% |
Molecular Formula | C6H15NO3 |
Quantity | 1 kg |
InChI Key | GSEJCLTVZPLZKY-UHFFFAOYSA-N |
Molecular Weight (g/mol) | 149.19 |
Formula Weight | 185.65 |
Physical Form | Powder |
Chemical Name or Material | Triethanolamine Hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'