Få mere at vide
Tridihexethyl Chloride, Mikromol™
Beskrivelse
Tridihexethyl Chloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tridihexethyl Chloride |
| CAS | 4310-35-4 |
| Analyt- eller komponentnavne | Tridihexethyl Chloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H36 N O . Cl |
| Certificeringer/compliance | ISO 17025 |
| Synonym | (3-Cyclohexyl-3-hydroxy-3-phenylpropyl)triethylammonium chloride (7CI), Ammonium, (3-cyclohexyl-3-hydroxy-3-phenylpropyl)triethyl-, chloride (8CI), 3-Diethylamino-1-cyclohexyl-1-phenyl-1-propanol-ethochloride, Pathilon chloride, Tridihexethyl chloride, Benzenepropanaminium, gamma-cyclohexyl-N,N,N-triethyl-gamma-hydroxy-, chloride (1:1), Benzenepropanaminium, gamma-cyclohexyl-N,N,N-triethyl-gamma-hydroxy-, chloride (9CI) |
| InChI formel | InChI=1S/C21H36NO.ClH/c1-4-22(5-2,6-3)18-17-21(23,19-13-9-7-10-14-19)20-15-11-8-12-16-20;/h7,9-10,13-14,20,23H,4-6,8,11-12,15-18H2,1-3H3;1H/q+1;/p-1 |
| SMIL | [Cl-].CC[N+](CC)(CC)CCC(O)(C1CCCCC1)c2ccccc2 |
| IUPAC navn | (3-cyclohexyl-3-hydroxy-3-phenylpropyl)-triethylazanium;chloride |
| Vis mere |