Få mere at vide
Trideuteromethyl Methanethiosulfonate, TRC
Artikelnummer. 30482752
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482752 25 mg
30468453 5 mg
30476387 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30482752 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32150325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 55800-37-8
Molekylær formel C2H3D3O2S2
Molekylvægt (g/mol) 129.22
Synonym Methanesulfonothioic Acid S-(Methyl-d3) Ester, Thiomethanesulfonic Acid S-(Methyl-d3) Ester, MMTS-d3, NSC 21545-d3, S-(Methyl-d3) Methanesulfonothioate, S-(Methyl-d3) Methanethiosulfonate
SMIL CCOC(=O)\C(C)=C(C)/N
Tekniske data
Kemisk navn eller materiale Trideuteromethyl Methanethiosulfonate