Få mere at vide
Tridecanoic-d25 Acid, TRC
Artikelnummer. 30452557
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30452557 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452557 Leverandør Toronto Research Chemicals Leverandørnr. TRCT774687100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tridecanoic-d25 Acid
CAS 202529-03-1
Molekylær formel C13 D25 H O2
Mængde 100 mg
Synonym NSC 25955-d25, NSC 69131-d25, Tridecylic-d25 Acid, n-Tridecanoic-d25 Acid, n-Tridecoic-d25 Acid, Tridecanoic-d25 acid (9CI), Tridecanoic acid-d25, Perdeuterotridecylic acid, Tridecanoic-D25 acid
InChI formel InChI=1S/C13H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13(14)15/h2-12H2,1H3,(H,14,15)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,13-pentacosadeuteriotridecanoic acid
Molekylvægt (g/mol) 239.5
Formel vægt 239.3502