missing translation for 'onlineSavingsMsg'
Få mere at vide

Tridecanoic-2,2-d2 Acid, CDN

Product Code. 30321471
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Mængde:
0.5 g
1 g
missing translation for 'unitSize'
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
30321471 0.5 g 0.5g
30309149 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321471 missing translation for 'mfr' CDN missing translation for 'supplierNo' CDND34710.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Tridecanoic-2,2-D2 Acid
CAS 64118-44-1
Molekylær formel C13 D2 H24 O2
Mængde 0.5 g
Synonym Tridecanoic-2,2-d2 acid (9 CI), Tridecanoic acid-d2, Tridecylic-α,α-d2 acid, Tridecanoic-2,2-D2 acid
InChI formel InChI=1 S/C13H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13(14)15/h2-12H2,1H3,(H,14,15)/i12D2
SMIL [2 H]C([2 H])(CCCCCCCCCCC)C(=O)O
IUPAC navn 2,2-dideuteriotridecanoic acid
Molekylvægt (g/mol) 216.357
Formel vægt 216.206 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.