Få mere at vide
Tridecanoic-2,2-d2 Acid, CDN
Artikelnummer. 30309149
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30309149 1 g -
30321471 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309149 Leverandør CDN Leverandørnr. CDND34711G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tridecanoic-2,2-D2 Acid
CAS 64118-44-1
Molekylær formel C13 D2 H24 O2
Mængde 1 g
Synonym Tridecanoic-2,2-d2 acid (9 CI), Tridecanoic acid-d2, Tridecylic-α,α-d2 acid, Tridecanoic-2,2-D2 acid
InChI formel InChI=1 S/C13H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13(14)15/h2-12H2,1H3,(H,14,15)/i12D2
SMIL [2 H]C([2 H])(CCCCCCCCCCC)C(=O)O
IUPAC navn 2,2-dideuteriotridecanoic acid
Molekylvægt (g/mol) 216.357
Formel vægt 216.206 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity