missing translation for 'onlineSavingsMsg'
Få mere at vide

Tridecafluorohexylethyl Methacrylate, TRC

Artikelnummer. 30452042
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30452042 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30452042 Leverandør Toronto Research Chemicals Leverandørnr. TRCT77507825G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Tridecafluorohexylethyl Methacrylate
CAS 2144-53-8
Renhedsgrad noter GC
Molekylær formel C12 H9 F13 O2
Mængde 25 g
Synonym 1H,1H,2H,2H-Perfluorooctyl Methacrylate, 2-Propenoic acid','2-methyl-','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl ester (9CI','ACI), 1-Octanol','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-','methacrylate (8CI), Methacrylic acid','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl ester (7CI','8CI), (Perfluorohexyl)ethyl methacrylate, 1H,1H,2H,2H-Perfluorooctyl methacrylate, 1H,1H,2H,2H-Tridecafluoro-1-octyl methacrylate, 1H,1H,2H,2H-Tridecafluorooctyl methacrylate, 1H,1H,2H,2H-Tridecafluorooctyl methacrylate, 2-(Perfluorohexyl)ethyl methacrylate, 2-(Tridecafluorohexyl)ethyl methacrylate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-yl 2-methyl-2-propenoate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctanyl methacrylate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl 2-methylprop-2-enoate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl methacrylate, 6:2 FTMAC, Actyflon G 06B, C 6FMA, C 6SFMA, Capstone 62MA, Cheminox FAMAC 6, FAMAC 6, Fluowet MA 600, M 1620, MA 600, MAC 6F13, Tridecafluorohexylethyl methacrylate, 6:2 FTMAC, 1H,1H,2H,2H-Perfluorooctyl Methacrylate, 2-Propenoic acid','2-methyl-','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl ester (9CI','ACI), 1-Octanol','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-','methacrylate (8CI), Methacrylic acid','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl ester (7CI','8CI), (Perfluorohexyl)ethyl methacrylate, 1H,1H,2H,2H-Perfluorooctyl methacrylate, 1H,1H,2H,2H-Tridecafluoro-1-octyl methacrylate, 1H,1H,2H,2H-Tridecafluorooctyl methacrylate, 1H,1H,2H,2H-Tridecafluorooctyl methacrylate, 2-(Perfluorohexyl)ethyl methacrylate, 2-(Tridecafluorohexyl)ethyl methacrylate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-yl 2-methyl-2-propenoate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctanyl methacrylate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl 2-methylprop-2-enoate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl methacrylate, 6:2 FTMAC, Actyflon G 06B, C 6FMA, C 6SFMA, Capstone 62MA, Cheminox FAMAC 6, FAMAC 6, Fluowet MA 600, M 1620, MA 600, MAC 6F13
InChI formel InChI=1S/C12H9F13O2/c1-5(2)6(26)27-4-3-7(13,14)8(15,16)9(17,18)10(19,20)11(21,22)12(23,24)25/h1,3-4H2,2H3
SMIL CC(=C)C(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC navn 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl 2-methylprop-2-enoate
Molekylvægt (g/mol) 432.18
Formel vægt 432.0395
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.