Få mere at vide
Euro Protection™ Tricot Seamless Knit Gloves
Artikelnummer. 12048195
:
Size 9
Artikelnummer. 12048195
Brand: Euro Protection™ 4365
Euro Protection™ Tricot Seamless Knit Gloves are comfortable and provide good grip and thermal contact insulation.
  • Anti-slip pvc dots on palm
  • Seamless, white cotton knit
  • Thermal contact insulation
  • Dexterity
  • Comfort
  • Ambidextrous
  • Suitable for transport and storage activities, handling of slippery objects, precise work
Tekniske data
Certificeringer/overholdelse (Europa) EN388:2016 012X, EN420
Belagt område Palm
Konstruktion Seamless Knit
Belægningsmateriale PVC Dots
Farve White
Hånd Ambidextrous
Materiale Cotton
Størrelse 9
Mængde 10/Pk