Få mere at vide
Triclocarban-d4 (4-chlorophenyl-d4), TRC
Artikelnummer. 30453886
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30453886 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453886 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7742015MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Triclocarban 4-chlorophenyl-d4
CAS 1219799-29-7
Renhedsgrad noter HPLC
Molekylær formel C13 2H4 H5 Cl3 N2 O
Mængde 5 mg
Synonym 3,4,4'-Trichlorocarbanilide-2',3',5',6'-d4, 3-(4-Chlorophenyl-2,3,5,6-d4)-1-(3,4-dichlorophenyl)urea, Triclocarban (4-chlorophenyl-D4)
InChI formel InChI=1S/C13H9Cl3N2O/c14-8-1-3-9(4-2-8)17-13(19)18-10-5-6-11(15)12(16)7-10/h1-7H,(H2,17,18,19)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(NC(=O)Nc2ccc(Cl)c(Cl)c2)c([2H])c([2H])c1Cl
IUPAC navn 1-(4-chloro-2,3,5,6-tetradeuteriophenyl)-3-(3,4-dichlorophenyl)urea
Molekylvægt (g/mol) 319.61
Formel vægt 318.0032
Procent renhed >95