missing translation for 'onlineSavingsMsg'
Få mere at vide

Triclocarban-d4 (4-chlorophenyl-d4), CDN

Artikelnummer. 30320425
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320425 0.05 g
30316204 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320425 Leverandør CDN Leverandørnr. CDND70290.05G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Triclocarban 4-chlorophenyl-d4
CAS 1219799-29-7
Molekylær formel C13 2H4 H5 Cl3 N2 O
Mængde 0.05 g
Synonym 3,4,4'-Trichlorocarbanilide-2',3',5',6'-d4, 3-(4-Chlorophenyl-2,3,5,6-d4)-1-(3,4-dichlorophenyl)urea, Triclocarban (4-chlorophenyl-D4)
InChI formel InChI=1 S/C13H9Cl3N2O/c14-8-1-3-9(4-2-8)17-13(19)18-10-5-6-11(15)12(16)7-10/h1-7 H,(H2,17,18,19)/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(NC(=O)Nc2ccc(Cl)c(Cl)c2)c([2 H])c([2 H])c1Cl
IUPAC navn 1-(4-chloro-2,3,5,6-tetradeuteriophenyl)-3-(3,4-dichlorophenyl)urea
Molekylvægt (g/mol) 319.61
Formel vægt 318.0032 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.