missing translation for 'onlineSavingsMsg'
Learn More

Tricine-buffered saline (5X), pH 8.0, Thermo Scientific™

Catalog Number p-7041926
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy
5704-04-1
C6H13NO5
179.17
MFCD00004277
SEQKRHFRPICQDD-UHFFFAOYSA-N
79784
CHEBI:39063
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
C(C(=O)O)NC(CO)(CO)CO
This item is not returnable. View return policy
5704-04-1
C6H13NO5
179.17
MFCD00004277
SEQKRHFRPICQDD-UHFFFAOYSA-N
79784
CHEBI:39063
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
C(C(=O)O)NC(CO)(CO)CO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Concentration 5X
pH 8.0
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material Tricine-buffered saline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'