Få mere at vide
Trichloronate, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Trichloronate |
| CAS | 327-98-0 |
| Analyt- eller komponentnavne | Trichloronate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C10 H12 Cl3 O2 P S |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Phosphonothioic acid, ethyl-, O-ethyl O-(2,4,5-trichlorophenyl) ester (8CI, 9CI, ACI), (±)-Trichloronate, 5082A, Agritox, BAY 37289, Bayer 37289, Bayer 5081, Bayer S 4400, Chemagro 37289, ENT-25712, Fitosol, O-Ethyl O-2,4,5-trichlorophenyl ethylphosphonothioate, Phytosol, Richloronate, S 4400, Trichloronat, Trichloronate |
| InChI formel | InChI=1S/C10H12Cl3O2PS/c1-3-14-16(17,4-2)15-10-6-8(12)7(11)5-9(10)13/h5-6H,3-4H2,1-2H3 |
| Løsningstype | Neat |
| SMIL | CCOP(=S)(CC)Oc1cc(Cl)c(Cl)cc1Cl |
| Vis mere |