missing translation for 'onlineSavingsMsg'
Få mere at vide

Trichloroethylene-d, CDN

Artikelnummer. 30325224
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325224 1 g
30316063 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325224 Leverandør CDN Leverandørnr. CDND39001G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Trichloroethylene-d
CAS 13291-68-4
Molekylær formel C2 D Cl3
Mængde 1 g
Synonym Ethene-d, trichloro- (9 CI), Ethylene-d, trichloro- (6 CI, 7 CI, 8 CI), 1,1,2-Trichloro-2-deuterioethene, Deuterotrichloroethylene
InChI formel InChI=1 S/C2HCl3/c3-1-2(4)5/h1H/i1D
SMIL [2 H]C(=C(Cl)Cl)Cl
IUPAC navn 1,1,2-trichloro-2-deuterioethene
Molekylvægt (g/mol) 132.3945
Formel vægt 130.9207 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity
Vis mere Vis mindre
Produkttitel
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.