Få mere at vide
Tributyl O-Acetylcitrate-d3, TRC
Artikelnummer. 30452218
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30452218 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452218 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7729525MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tributyl O-Acetylcitrate-d3
CAS 1794753-49-3
Molekylær formel C20D3H31O8
Mængde 5 mg
Synonym 2-(Acetyloxy-d3)-1,2,3-propanetricarboxylic Acid 1,2,3-Tributyl Ester, Citric Acid Tributyl Ester Acetate-d3, ATBC-d3, Acetyl-d3 Butyl Citrate, Acetyl-d3 Tributyl Citrate, (Acetyl-d3)citric Acid Tributyl Ester, Blo-trol-d3, Citroflex 4A-S-d3, Citroflex A-d3, Citroflex A 4-d3, Citrofol BII-d3, Estaflex ATC-d3, Monocizer ATBC-d3, Morflex ATBC-d3, NSC 3894, Sansocizer ATBC, Tributyl 2-(Acetoxy-d3)-1,2,3-propane-tricarboxylate, Tributyl (Acetyl-d3)citrate, Tributyl Citrate Acetate-d3, ATBC-d3
InChI formel InChI=1S/C20H34O8/c1-5-8-11-25-17(22)14-20(28-16(4)21,19(24)27-13-10-7-3)15-18(23)26-12-9-6-2/h5-15H2,1-4H3/i4D3
SMIL [2H]C([2H])([2H])C(=O)OC(CC(=O)OCCCC)(CC(=O)OCCCC)C(=O)OCCCC
IUPAC navn tributyl 2-(2,2,2-trideuterioacetyl)oxypropane-1,2,3-tricarboxylate
Molekylvægt (g/mol) 405.5
Formel vægt 405.2442