Få mere at vide
Tribenzothiophene, TRC
Artikelnummer. 30450150
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30450150 100 mg
30452961 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30450150 Leverandør Toronto Research Chemicals Leverandørnr. TRCT767850100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 30796-92-0
Molekylær formel C14H8S
Molekylvægt (g/mol) 208.28
Synonym 4,5-Epithiophenanthrene, Benzo[def]dibenzothiophene, Phenanthro[4,5-bcd]thiophene
SMIL c1cc2ccc3cccc4sc(c1)c2c34
Tekniske data
Kemisk navn eller materiale Tribenzothiophene
Mængde 100 mg
InChI formel InChI=1S/C14H8S/c1-3-9-7-8-10-4-2-6-12-14(10)13(9)11(5-1)15-12/h1-8H
Formel vægt 208.0347