Få mere at vide
Triazophos-D5, TRC
Artikelnummer. 30452755
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30452755 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452755 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7675925MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Triazophos D5 (Phenyl D5)
CAS 1773496-62-0
Molekylær formel C12 2H5 H11 N3 O3 P S
Mængde 5 mg
Synonym Triazophos D5 (Phenyl D5)
InChI formel InChI=1S/C12H16N3O3PS/c1-3-16-19(20,17-4-2)18-12-13-10-15(14-12)11-8-6-5-7-9-11/h5-10H,3-4H2,1-2H3/i5D,6D,7D,8D,9D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])n2cnc(OP(=S)(OCC)OCC)n2
IUPAC navn diethoxy-[[1-(2,3,4,5,6-pentadeuteriophenyl)-1,2,4-triazol-3-yl]oxy]-sulfanylidene-λ^{5}-phosphane
Molekylvægt (g/mol) 318.34
Formel vægt 318.0964