Få mere at vide
Triamterene-d5, TRC
Artikelnummer. 30449193
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30449193 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449193 Leverandør Toronto Research Chemicals Leverandørnr. TRCT76732710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Triamterene-d5
CAS 1189922-23-3
Molekylær formel C12 D5 H6 N7
Mængde 10 mg
Synonym Triamterene D5 (ring D5), Triamterene D5 (phenyl D5), 6-(2,3,4,5,6-Pentadeuteriophenyl)pteridine-2,4,7-triamine
InChI formel InChI=1S/C12H11N7/c13-9-7(6-4-2-1-3-5-6)16-8-10(14)18-12(15)19-11(8)17-9/h1-5H,(H6,13,14,15,17,18,19)/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2nc3c(N)nc(N)nc3nc2N
IUPAC navn 6-(2,3,4,5,6-pentadeuteriophenyl)pteridine-2,4,7-triamine
Molekylvægt (g/mol) 258.293
Formel vægt 258.139