Få mere at vide
Triamcinolone Hexacetonide, LoGiCal
Artikelnummer. 30462697
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462697 10 mg
1 options
Artikelnummer. 30462697 Leverandør LoGiCal Leverandørnr. LGCFOR0306.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Triamcinolone Hexacetonide
CAS 5611-51-8
Analyt- eller komponentnavne Triamcinolone Hexacetonide
Forsendelsestilstand Room Temperature
Molekylær formel C30 H41 F O7
Certificeringer/compliance ISO 9001
Synonym Triamcinolone Hexacetonide, Aristospan, CL 34433, Hexatrione, Lederlon, Lederspan, TATBA, Triamcinolone acetonide 21-tert-butylacetate, Triamcinolone acetonide tert-butyl acetate, Pregna-1,4-diene-3,20-dione, 21-(3,3-dimethyl-1-oxobutoxy)-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (11beta,16alpha)-, Pregna-1,4-diene-3,20-dione, 9-fluoro-11beta,16alpha,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone, 21-(3,3-dimethylbutyrate) (8CI), (11beta,16alpha)-21-(3,3-Dimethyl-1-oxobutoxy)-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, Butyric acid, 3,3-dimethyl-, 21-ester with 9-fluoro-11beta,16alpha,17,21-tetrahydroxypregna-1,4-diene-3,20-dione cyclic 16,17-acetal with acetone (8CI)
InChI formel InChI=1S/C30H41FO7/c1-25(2,3)15-24(35)36-16-22(34)30-23(37-26(4,5)38-30)13-20-19-9-8-17-12-18(32)10-11-27(17,6)29(19,31)21(33)14-28(20,30)7/h10-12,19-21,23,33H,8-9,13-16H2,1-7H3/t19-,20-,21-,23+,27-,28-,29-,30+/m0/s1
Løsningstype Neat
SMIL CC(C)(C)CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]23C
IUPAC navn (11β,16α)-21-(3,3-Dimethyl-1-oxobutoxy)-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione
Industry Type Clinical Testing
Molekylvægt (g/mol) 532.64
Mængde 10 mg
Formel vægt 532.2836 g/mol