Få mere at vide
Triamcinolone Furetonide, TRC
Artikelnummer. 30456408
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456408 250 mg
30451167 50 mg
30452716 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30456408 Leverandør Toronto Research Chemicals Leverandørnr. TRCT720860250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4989-94-0
Molekylær formel C33H35FO8
Molekylvægt (g/mol) 578.62
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11beta,16alpha,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone, 21-(2-benzofurancarboxylate), (11beta,16alpha)-21-[(2-Benzofuranylcarbonyl)oxy]-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, Triamcinolone Acetonide Coumarilic Acid Ester
SMIL CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)COC(=O)c6oc7ccccc7c6
Tekniske data
Kemisk navn eller materiale Triamcinolone Furetonide
Mængde 250 mg
InChI formel InChI=1S/C33H35FO8/c1-29(2)41-27-15-22-21-10-9-19-14-20(35)11-12-30(19,3)32(21,34)25(36)16-31(22,4)33(27,42-29)26(37)17-39-28(38)24-13-18-7-5-6-8-23(18)40-24/h5-8,11-14,21-22,25,27,36H,9-10,15-17H2,1-4H3/t21-,22-,25-,27+,30-,31-,32-,33+/m0/s1
Formel vægt 578.2316