Få mere at vide
Triamcinolone Acetonide, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Triamcinolone acetonide |
| CAS | 76-25-5 |
| Analyt- eller komponentnavne | Triamcinolone acetonide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C24 H31 F O6 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Triamcinolone Hexacetonide Imp. A (EP), Triamcinolone Acetonide, Pregna-1,4-diene-3,20-dione, 9-fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (11beta,16alpha)-, Pregna-1,4-diene-3,20-dione, 9-fluoro-11beta,16alpha,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone (6CI,8CI), (11beta,16alpha)-9-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, 9-Fluoro-11beta,16alpha,17,21-tetrahydroxypregna-1,4-diene-3,20-dione-16,17-acetonide, 9-Fluoro-11beta,21-dihydroxy-16alpha,17-(isopropylidenedioxy)pregna-1,4-diene-3,20-dione, 9-Fluoro-11beta,21-dihydroxy-16alpha,17alpha-(isopropylidenedioxy)pregna-1,4-diene-3,20-dione, 9alpha-Fluoro-11beta,21-dihydroxy-16alpha,17alpha-isopropylidenedioxy-1,4-pregnadiene-3,20-dione, 9alpha-Fluoro-16alpha-hydroxyprednisolone 16alpha,17alpha-acetonide, Adcortyl, Adcortyl A, Aftach, Aristicort, Aristocort A, Aristocort acetonide, Aristoderm, Azmacort, Berlicort, Delphicort, Extracort, Ftorocort, I-Vation, Kenacort 40, Kenacort 80, Kenacort A, Kenacort A 40, Kenalog, Kenalog 10, Kenalog 40, Ledercort Cream, Lichtena, MaQuaid, NSC 21916, Nasacort, Omcilon A, Paralen, Pol'kortolon, Queyanshusong A, Respicort, Rheudenolone, Rineton, Solodelf, TAC 3, TAC 40, Tamceton, Tramacin, Tri-Nasal, Triacet, Triakort, Triam, Triamcinolone 16,17-acetonide, Triamcinolone 16alpha,17alpha-acetonide, Triamcort-Depot, Triamhexal, Triamonide 40, Trianopollon, Tricinolon, Triesence, Trivaris, Trymex, Valon A 10, Vetalog, Vetazine, Volon A, Volonimat, 9-Fluoro-11beta,21-dihydroxy-16alpha,17-(1-methylethylidenedioxy)pregna-1,4-diene-3,20-dione, Fluocinolone Acetonide Impurity H, Triamcinolone Hexacetonide Impurity A |
| InChI formel | InChI=1S/C24H31FO6/c1-20(2)30-19-10-16-15-6-5-13-9-14(27)7-8-21(13,3)23(15,25)17(28)11-22(16,4)24(19,31-20)18(29)12-26/h7-9,15-17,19,26,28H,5-6,10-12H2,1-4H3/t15-,16-,17-,19+,21-,22-,23-,24+/m0/s1 |
| Løsningstype | Neat |
| SMIL | CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO |
| Vis mere |